Product Details
- SNP ID
-
rs144545876
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:89326831 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGGTTACTCAGTTTATGAAGAGT[C/T]TTGGATAGTGAATTGGGTCCACAAC
- Phenotype
-
MIM: 605261
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NOX4
PubMed Links
Gene Details
- Gene
- NOX4
- Gene Name
- NADPH oxidase 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001143836.2 |
1828 |
Silent Mutation |
AAA,AAG |
K514K |
NP_001137308.1 |
NM_001143837.1 |
1828 |
Intron |
|
|
NP_001137309.1 |
NM_001291926.1 |
1828 |
Silent Mutation |
AAA,AAG |
K480K |
NP_001278855.1 |
NM_001291927.1 |
1828 |
Silent Mutation |
AAA,AAG |
K575K |
NP_001278856.1 |
NM_001291929.1 |
1828 |
Silent Mutation |
AAA,AAG |
K529K |
NP_001278858.1 |
NM_001300995.1 |
1828 |
Silent Mutation |
AAA,AAG |
K490K |
NP_001287924.1 |
NM_016931.4 |
1828 |
Silent Mutation |
AAA,AAG |
K554K |
NP_058627.1 |
XM_006718849.3 |
1828 |
Silent Mutation |
AAA,AAG |
K535K |
XP_006718912.1 |
XM_011542857.2 |
1828 |
Silent Mutation |
AAA,AAG |
K530K |
XP_011541159.1 |
XM_017017841.1 |
1828 |
Silent Mutation |
AAA,AAG |
K548K |
XP_016873330.1 |
XM_017017842.1 |
1828 |
Intron |
|
|
XP_016873331.1 |
XM_017017843.1 |
1828 |
Intron |
|
|
XP_016873332.1 |
XM_017017844.1 |
1828 |
Silent Mutation |
AAA,AAG |
K388K |
XP_016873333.1 |
XM_017017845.1 |
1828 |
Silent Mutation |
AAA,AAG |
K388K |
XP_016873334.1 |
View Full Product Details