Product Details
- SNP ID
-
rs145145100
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:61411920 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAATGGCTGTGAGCAGCGTCTCAA[C/T]TGCGTCACTGTAATCCCCTGATAAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CPSF7
PubMed Links
Gene Details
- Gene
- CPSF7
- Gene Name
- cleavage and polyadenylation specific factor 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001136040.2 |
1313 |
Missense Mutation |
ATT,GTT |
I368V |
NP_001129512.1 |
NM_001142565.1 |
1313 |
Missense Mutation |
ATT,GTT |
I359V |
NP_001136037.1 |
NM_024811.3 |
1313 |
Missense Mutation |
ATT,GTT |
I411V |
NP_079087.3 |
XM_005274298.4 |
1313 |
Missense Mutation |
ATT,GTT |
I402V |
XP_005274355.1 |
XM_005274299.4 |
1313 |
Missense Mutation |
ATT,GTT |
I368V |
XP_005274356.1 |
XM_005274303.4 |
1313 |
Missense Mutation |
ATT,GTT |
I278V |
XP_005274360.1 |
XM_011545257.2 |
1313 |
Missense Mutation |
ATT,GTT |
I344V |
XP_011543559.1 |
XM_011545258.2 |
1313 |
Missense Mutation |
ATT,GTT |
I368V |
XP_011543560.1 |
XM_011545259.2 |
1313 |
Missense Mutation |
ATT,GTT |
I359V |
XP_011543561.1 |
XM_011545260.2 |
1313 |
Missense Mutation |
ATT,GTT |
I278V |
XP_011543562.1 |
XM_011545261.2 |
1313 |
Missense Mutation |
ATT,GTT |
I278V |
XP_011543563.1 |
XM_011545262.2 |
1313 |
Missense Mutation |
ATT,GTT |
I134V |
XP_011543564.1 |
XM_011545263.2 |
1313 |
Missense Mutation |
ATT,GTT |
I134V |
XP_011543565.1 |
XM_017018345.1 |
1313 |
Missense Mutation |
ATT,GTT |
I335V |
XP_016873834.1 |
View Full Product Details