Product Details

SNP ID
rs145372004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56176912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAATTTCTTTCTCTGCTTGCATG[G/T]TACAGCATTTGTGTTTCGGAGTGTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR5J2 PubMed Links

Gene Details

Gene
OR5J2
Gene Name
olfactory receptor family 5 subfamily J member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005492.1 295 Missense Mutation GTA,TTA V99L NP_001005492.1

View Full Product Details