Product Details

SNP ID
rs145373332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6999884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTATAAGTTTCCTCTTCTATGA[C/T]TATTGTGAAGATGTGAATTATGATC
Phenotype
MIM: 605015
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF214 PubMed Links

Gene Details

Gene
ZNF214
Gene Name
zinc finger protein 214
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013249.2 2177 Missense Mutation AAT,AGT N600S NP_037381.2
XM_005253128.3 2177 Missense Mutation AAT,AGT N600S XP_005253185.1
XM_005253129.3 2177 Missense Mutation AAT,AGT N586S XP_005253186.1
XM_006718308.3 2177 Missense Mutation AAT,AGT N600S XP_006718371.1
XM_006718309.3 2177 Missense Mutation AAT,AGT N586S XP_006718372.1
XM_011520355.2 2177 Missense Mutation AAT,AGT N611S XP_011518657.1
XM_011520357.2 2177 Missense Mutation AAT,AGT N586S XP_011518659.1
XM_017018269.1 2177 Missense Mutation AAT,AGT N586S XP_016873758.1

View Full Product Details