Product Details

SNP ID
rs145545312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:48264156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCAAGGGCTGCCTCACACAGA[C/T]GTTTTCCCTCCATTTCTTTGGTGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4X1 PubMed Links

Gene Details

Gene
OR4X1
Gene Name
olfactory receptor family 4 subfamily X member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004726.1 296 Missense Mutation ACG,ATG T99M NP_001004726.1

View Full Product Details