Product Details

SNP ID
rs145860209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:36592995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAATTCATCATCACCATCAAAAC[C/T]ATTTGCTTCTGCACTGAAACAAAAT
Phenotype
MIM: 179616
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf74 PubMed Links

Gene Details

Gene
C11orf74
Gene Name
chromosome 11 open reading frame 74
There are no transcripts associated with this gene.

Gene
RAG2
Gene Name
recombination activating gene 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000536.3 1379 Missense Mutation AGT,GGT S392G NP_000527.2
NM_001243785.1 1379 Missense Mutation AGT,GGT S392G NP_001230714.1
NM_001243786.1 1379 Missense Mutation AGT,GGT S392G NP_001230715.1

View Full Product Details