Product Details

SNP ID
rs145892725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55795926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTGGCAGTGATGGCCTATGACC[A/G]CTTTGTGGCCATCTGCAATCCTCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5D14 PubMed Links

Gene Details

Gene
OR5D14
Gene Name
olfactory receptor family 5 subfamily D member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004735.1 371 Missense Mutation CAC,CGC H124R NP_001004735.1

View Full Product Details