Product Details

SNP ID
rs145926656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111883827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCAACCTGAACTGGATCCTCCC[C/T]GATACAAATGCACAGAAAAGTCAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf1 PubMed Links

Gene Details

Gene
C11orf1
Gene Name
chromosome 11 open reading frame 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022761.2 506 Nonsense Mutation CGA,TGA R134* NP_073598.1
XM_011542957.2 506 Nonsense Mutation CGA,TGA R150* XP_011541259.1
XM_011542958.2 506 Silent Mutation CCC,CCT P127P XP_011541260.1
XM_017018129.1 506 Silent Mutation CCC,CCT P111P XP_016873618.1
Gene
FDXACB1
Gene Name
ferredoxin-fold anticodon binding domain containing 1
There are no transcripts associated with this gene.

View Full Product Details