Product Details

SNP ID
rs145976732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67451377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGGGGCTTTCTCCTTCAGAGG[C/T]AGGAGGTGTGGCTGGGTCCTCAAGG
Phenotype
MIM: 608965 MIM: 609849
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABP4 PubMed Links

Gene Details

Gene
CABP4
Gene Name
calcium binding protein 4
There are no transcripts associated with this gene.

Gene
CORO1B
Gene Name
coronin 1B
There are no transcripts associated with this gene.

Gene
GPR152
Gene Name
G protein-coupled receptor 152
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206997.1 1353 Missense Mutation ACC,GCC T450A NP_996880.1

View Full Product Details