Product Details
- SNP ID
-
rs146137167
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:96353704 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCAAAATTGAGATATTCTTCAAGT[C/T]GACCATATTTCTGCATATACAATAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC82
PubMed Links
Gene Details
- Gene
- CCDC82
- Gene Name
- coiled-coil domain containing 82
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318736.1 |
1926 |
Missense Mutation |
CAA,CGA |
Q526R |
NP_001305665.1 |
NM_001318737.1 |
1926 |
Intron |
|
|
NP_001305666.1 |
NM_024725.3 |
1926 |
Missense Mutation |
CAA,CGA |
Q526R |
NP_079001.2 |
XM_005274292.3 |
1926 |
Missense Mutation |
CAA,CGA |
Q526R |
XP_005274349.1 |
XM_006718911.3 |
1926 |
Missense Mutation |
CAA,CGA |
Q503R |
XP_006718974.1 |
XM_011542984.2 |
1926 |
Missense Mutation |
CAA,CGA |
Q526R |
XP_011541286.1 |
XM_011542987.2 |
1926 |
Missense Mutation |
CAA,CGA |
Q526R |
XP_011541289.1 |
XM_011542988.2 |
1926 |
Missense Mutation |
CAA,CGA |
Q526R |
XP_011541290.1 |
XM_017018305.1 |
1926 |
Missense Mutation |
CAA,CGA |
Q526R |
XP_016873794.1 |
XM_017018306.1 |
1926 |
Missense Mutation |
CAA,CGA |
Q526R |
XP_016873795.1 |
XM_017018307.1 |
1926 |
Missense Mutation |
CAA,CGA |
Q526R |
XP_016873796.1 |
XM_017018308.1 |
1926 |
Missense Mutation |
CAA,CGA |
Q526R |
XP_016873797.1 |
XM_017018309.1 |
1926 |
UTR 3 |
|
|
XP_016873798.1 |
XM_017018310.1 |
1926 |
UTR 3 |
|
|
XP_016873799.1 |
XM_017018311.1 |
1926 |
Missense Mutation |
CAA,CGA |
Q503R |
XP_016873800.1 |
XM_017018312.1 |
1926 |
Missense Mutation |
CAA,CGA |
Q503R |
XP_016873801.1 |
View Full Product Details