Product Details

SNP ID
rs146894544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124570504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACCGCTATGTTGCCATCTGCCAC[C/G]CTTTGCTTTACAACATCATTATGTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR8A1 PubMed Links

Gene Details

Gene
OR8A1
Gene Name
olfactory receptor family 8 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005194.1 436 Missense Mutation CCT,GCT P146A NP_001005194.1

View Full Product Details