Product Details

SNP ID
rs147652295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65501589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGTTTTTTTCTAAGATTTTCC[A/G]CAGATGCTATAGTACTATTGACAAA
Phenotype
MIM: 607924
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MALAT1 PubMed Links

Gene Details

Gene
MALAT1
Gene Name
metastasis associated lung adenocarcinoma transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details