Product Details

SNP ID
rs147846695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5777914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTGAAAGCCTTCTGCCGAGCAT[C/G]TGATGAAGAGAGGCTGATCGCTGCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR52N1 PubMed Links

Gene Details

Gene
OR52N1
Gene Name
olfactory receptor family 52 subfamily N member 1
There are no transcripts associated with this gene.

Gene
OR52N5
Gene Name
olfactory receptor family 52 subfamily N member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001922.2 754 Missense Mutation CAT,GAT H241D NP_001001922.2

View Full Product Details