Product Details
- SNP ID
-
rs147877589
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:46379919 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCGGCAGCACTACCAGATGATCC[A/G]GCGGGAGGACCAGGAGACGGCTGTG
- Phenotype
-
MIM: 118495
MIM: 601441
MIM: 162096
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHRM4
PubMed Links
Gene Details
- Gene
- CHRM4
- Gene Name
- cholinergic receptor muscarinic 4
There are no transcripts associated with this gene.
- Gene
- DGKZ
- Gene Name
- diacylglycerol kinase zeta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001105540.1 |
2846 |
Missense Mutation |
CAG,CGG |
Q1109R |
NP_001099010.1 |
NM_001199266.1 |
2846 |
Missense Mutation |
CAG,CGG |
Q926R |
NP_001186195.1 |
NM_001199267.1 |
2846 |
Missense Mutation |
CAG,CGG |
Q920R |
NP_001186196.1 |
NM_001199268.1 |
2846 |
Missense Mutation |
CAG,CGG |
Q898R |
NP_001186197.1 |
NM_003646.3 |
2846 |
Missense Mutation |
CAG,CGG |
Q921R |
NP_003637.2 |
NM_201532.2 |
2846 |
Missense Mutation |
CAG,CGG |
Q937R |
NP_963290.1 |
NM_201533.3 |
2846 |
Missense Mutation |
CAG,CGG |
Q925R |
NP_963291.2 |
XM_005253181.3 |
2846 |
Missense Mutation |
CAG,CGG |
Q938R |
XP_005253238.1 |
XM_005253182.3 |
2846 |
Missense Mutation |
CAG,CGG |
Q926R |
XP_005253239.1 |
XM_006718354.1 |
2846 |
Missense Mutation |
CAG,CGG |
Q1110R |
XP_006718417.1 |
XM_006718355.2 |
2846 |
Missense Mutation |
CAG,CGG |
Q925R |
XP_006718418.1 |
XM_011520422.1 |
2846 |
Missense Mutation |
CAG,CGG |
Q903R |
XP_011518724.1 |
XM_011520423.1 |
2846 |
Missense Mutation |
CAG,CGG |
Q801R |
XP_011518725.1 |
XM_017018455.1 |
2846 |
Intron |
|
|
XP_016873944.1 |
XM_017018456.1 |
2846 |
Intron |
|
|
XP_016873945.1 |
XM_017018457.1 |
2846 |
Intron |
|
|
XP_016873946.1 |
- Gene
- MDK
- Gene Name
- midkine (neurite growth-promoting factor 2)
- Gene
- MIR4688
- Gene Name
- microRNA 4688
There are no transcripts associated with this gene.
View Full Product Details