Product Details
- SNP ID
-
rs148214991
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:118531833 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCCTCCAGGCCCAGCCGCCCTCC[A/G]GCACACACTGCTGCTCCTGCCAGCC
- Phenotype
-
MIM: 159555
MIM: 613934
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KMT2A
PubMed Links
Gene Details
- Gene
- KMT2A
- Gene Name
- lysine methyltransferase 2A
There are no transcripts associated with this gene.
- Gene
- LOC101929089
- Gene Name
- uncharacterized LOC101929089
There are no transcripts associated with this gene.
- Gene
- TMEM25
- Gene Name
- transmembrane protein 25
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144034.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
NP_001137506.1 |
NM_001144035.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
NP_001137507.1 |
NM_001144036.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
NP_001137508.1 |
NM_001144037.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
NP_001137509.1 |
NM_001144038.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
NP_001137510.1 |
NM_001318755.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
NP_001305684.1 |
NM_001318757.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
NP_001305686.1 |
NM_032780.3 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
NP_116169.2 |
XM_005271703.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_005271760.1 |
XM_006718926.2 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_006718989.1 |
XM_006718927.2 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_006718990.1 |
XM_011543037.2 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_011541339.1 |
XM_011543038.2 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_011541340.1 |
XM_017018422.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_016873911.1 |
XM_017018423.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_016873912.1 |
XM_017018424.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_016873913.1 |
XM_017018425.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_016873914.1 |
XM_017018426.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_016873915.1 |
XM_017018427.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_016873916.1 |
XM_017018428.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_016873917.1 |
XM_017018429.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_016873918.1 |
XM_017018430.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_016873919.1 |
XM_017018431.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_016873920.1 |
XM_017018432.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_016873921.1 |
XM_017018433.1 |
92 |
Missense Mutation |
CAG,CGG |
Q11R |
XP_016873922.1 |
- Gene
- TTC36
- Gene Name
- tetratricopeptide repeat domain 36
View Full Product Details