Product Details

SNP ID
rs148287677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68708485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCTCCACCACCTCCCAGGAGAT[A/G]CATGAGGAAGGCCGCCTGTCAGAAA
Phenotype
MIM: 604374
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TESMIN PubMed Links

Gene Details

Gene
TESMIN
Gene Name
testis expressed metallothionein like protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039656.1 1336 Intron NP_001034745.1
NM_004923.3 1336 Silent Mutation TGC,TGT C450C NP_004914.2
XM_011545402.1 1336 Silent Mutation TGC,TGT C450C XP_011543704.1
XM_011545403.2 1336 Silent Mutation TGC,TGT C450C XP_011543705.1
XM_017018588.1 1336 Silent Mutation TGC,TGT C384C XP_016874077.1

View Full Product Details