Product Details

SNP ID
rs148357349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7796196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCACATAAATGAAGGTAATGGT[C/T]CCATAGAACAGGGTAACCACAGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5P2 PubMed Links

Gene Details

Gene
OR5P2
Gene Name
olfactory receptor family 5 subfamily P member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153444.1 747 Silent Mutation GGA,GGG G249G NP_703145.1

View Full Product Details