Product Details

SNP ID
rs148799278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69005507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGACGTACTCGGGGAAGGGGGCC[A/G]GGATCTGGAAGACCCAGAAGAGGAA
Phenotype
MIM: 607233
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRGPRF PubMed Links

Gene Details

Gene
MRGPRF
Gene Name
MAS related GPR family member F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098515.1 1668 Missense Mutation CCG,CTG P268L NP_001091985.1
NM_145015.4 1668 Missense Mutation CCG,CTG P268L NP_659452.3
XM_017017170.1 1668 Missense Mutation CCG,CTG P268L XP_016872659.1
Gene
MRGPRF-AS1
Gene Name
MRGPRF antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details