Product Details

SNP ID
rs149054498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47420307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGATCTGCATGATTCTGGCCC[A/G]GGCCTCCTCATTGGGCATCGGGAAC
Phenotype
MIM: 186852 MIM: 608735
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMC3 PubMed Links

Gene Details

Gene
PSMC3
Gene Name
proteasome 26S subunit, ATPase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002804.4 1278 Missense Mutation CGG,TGG R362W NP_002795.2
XM_017018026.1 1278 Missense Mutation CGG,TGG R362W XP_016873515.1
Gene
SLC39A13
Gene Name
solute carrier family 39 member 13
There are no transcripts associated with this gene.

View Full Product Details