Product Details

SNP ID
rs149305921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63544875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCACCCAGCTGAGATATGGCAAG[G/T]CCCGCTGTAAACAGGTAAGGACCTC
Phenotype
MIM: 613866 MIM: 605092
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HRASLS2 PubMed Links

Gene Details

Gene
HRASLS2
Gene Name
HRAS like suppressor 2
There are no transcripts associated with this gene.

Gene
RARRES3
Gene Name
retinoic acid receptor responder 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004585.4 441 Missense Mutation GCC,TCC A125S NP_004576.2

View Full Product Details