Product Details

SNP ID
rs149541409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:30232033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTTCTGCATAAGCATCAACACCA[C/G]TTGGTGTGCTGGCTACTGCTACACC
Phenotype
MIM: 136530
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FSHB PubMed Links

Gene Details

Gene
FSHB
Gene Name
follicle stimulating hormone beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000510.2 200 Missense Mutation ACT,AGT T44S NP_000501.1
NM_001018080.1 200 Missense Mutation ACT,AGT T44S NP_001018090.1

View Full Product Details