Product Details

SNP ID
rs149670543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58507423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAAGACTTTCTTCTGCCCTTCA[C/G]CAGAATGCATCCTCTGAATGGTGAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR5B21 PubMed Links

Gene Details

Gene
OR5B21
Gene Name
olfactory receptor family 5 subfamily B member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005218.1 683 Missense Mutation GCT,GGT A228G NP_001005218.1

View Full Product Details