Product Details

SNP ID
rs149678247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:76680233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGAAAAGTGACGTCGCAAGGTCTG[C/T]GTTGCAGGTAGATTAGGGGTGGGAA
Phenotype
MIM: 601138 MIM: 137207
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCY2EP PubMed Links

Gene Details

Gene
GUCY2EP
Gene Name
guanylate cyclase 2E, pseudogene
There are no transcripts associated with this gene.

Gene
LRRC32
Gene Name
leucine rich repeat containing 32
There are no transcripts associated with this gene.

View Full Product Details