Product Details

SNP ID
rs149828052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57387780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGTACACAGGGCCACGCAGTGA[A/C]CACCGCGGGACCAGGGCTGGTGAGC
Phenotype
MIM: 605601 MIM: 606814
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRG2 PubMed Links

Gene Details

Gene
PRG2
Gene Name
proteoglycan 2, pro eosinophil major basic protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243245.2 771 Missense Mutation GGT,GTT G184V NP_001230174.1
NM_001302926.1 771 Missense Mutation GGT,GTT G195V NP_001289855.1
NM_001302927.1 771 Missense Mutation GGT,GTT G195V NP_001289856.1
NM_002728.5 771 Missense Mutation GGT,GTT G195V NP_002719.3
Gene
PRG3
Gene Name
proteoglycan 3, pro eosinophil major basic protein 2
There are no transcripts associated with this gene.

View Full Product Details