Product Details

SNP ID
rs149917532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7665401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGGTGAAAATCATGTCCTGGGT[A/G]TAACCCAGCTTCTCCAGGTTAGGGT
Phenotype
MIM: 608342 MIM: 603142
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYB5R2 PubMed Links

Gene Details

Gene
CYB5R2
Gene Name
cytochrome b5 reductase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302826.1 1027 Silent Mutation TAC,TAT Y268Y NP_001289755.1
NM_001302827.1 1027 UTR 3 NP_001289756.1
NM_016229.4 1027 Silent Mutation TAC,TAT Y268Y NP_057313.2
XM_005252975.4 1027 Silent Mutation TAC,TAT Y268Y XP_005253032.1
XM_006718251.2 1027 Silent Mutation TAC,TAT Y268Y XP_006718314.1
XM_011520184.2 1027 Silent Mutation TAC,TAT Y328Y XP_011518486.1
XM_011520185.2 1027 UTR 3 XP_011518487.1
XM_017017920.1 1027 Silent Mutation TAC,TAT Y328Y XP_016873409.1
XM_017017921.1 1027 UTR 3 XP_016873410.1
XM_017017922.1 1027 UTR 3 XP_016873411.1
Gene
PPFIBP2
Gene Name
PPFIA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256568.1 1027 Intron NP_001243497.1
NM_001256569.1 1027 Intron NP_001243498.1
NM_003621.3 1027 Intron NP_003612.2
XM_005253177.2 1027 Intron XP_005253234.1
XM_006718349.2 1027 Intron XP_006718412.1
XM_006718350.2 1027 Intron XP_006718413.1
XM_006718351.2 1027 Intron XP_006718414.1
XM_006718352.2 1027 Intron XP_006718415.1
XM_006718353.2 1027 Intron XP_006718416.1
XM_011520413.2 1027 Intron XP_011518715.1
XM_011520414.2 1027 Intron XP_011518716.1
XM_011520415.2 1027 Intron XP_011518717.1
XM_011520416.2 1027 Intron XP_011518718.1
XM_011520417.2 1027 Intron XP_011518719.1
XM_011520418.2 1027 Intron XP_011518720.1
XM_011520419.2 1027 Intron XP_011518721.1
XM_011520420.2 1027 Intron XP_011518722.1
XM_017018443.1 1027 Intron XP_016873932.1
XM_017018444.1 1027 Intron XP_016873933.1
XM_017018445.1 1027 Intron XP_016873934.1
XM_017018446.1 1027 Intron XP_016873935.1
XM_017018447.1 1027 Intron XP_016873936.1

View Full Product Details