Product Details
- SNP ID
-
rs150173703
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:772468 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTCGAACACCCAGGATCTGTCCT[C/T]GTTGGTGGCACAGCACAGGGCGGCG
- Phenotype
-
MIM: 602063
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOC171391
PubMed Links
Gene Details
- Gene
- LOC171391
- Gene Name
- uncharacterized LOC171391
There are no transcripts associated with this gene.
- Gene
- PDDC1
- Gene Name
- Parkinson disease 7 domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318818.1 |
434 |
Missense Mutation |
AAG,GAG |
K87E |
NP_001305747.1 |
NM_001318820.1 |
434 |
Missense Mutation |
AAG,GAG |
K101E |
NP_001305749.1 |
NM_001318821.1 |
434 |
Missense Mutation |
AAG,GAG |
K137E |
NP_001305750.1 |
NM_001318822.1 |
434 |
Missense Mutation |
AAG,GAG |
K51E |
NP_001305751.1 |
NM_001318823.1 |
434 |
Missense Mutation |
AAG,GAG |
K87E |
NP_001305752.1 |
NM_001318824.1 |
434 |
Missense Mutation |
AAG,GAG |
K101E |
NP_001305753.1 |
NM_182612.3 |
434 |
Missense Mutation |
AAG,GAG |
K137E |
NP_872418.1 |
XM_005252898.2 |
434 |
Missense Mutation |
AAG,GAG |
K137E |
XP_005252955.1 |
XM_011520064.1 |
434 |
Missense Mutation |
AAG,GAG |
K137E |
XP_011518366.1 |
XM_017017660.1 |
434 |
Missense Mutation |
AAG,GAG |
K137E |
XP_016873149.1 |
XM_017017661.1 |
434 |
Missense Mutation |
AAG,GAG |
K137E |
XP_016873150.1 |
XM_017017662.1 |
434 |
Missense Mutation |
AAG,GAG |
K137E |
XP_016873151.1 |
XM_017017663.1 |
434 |
Missense Mutation |
AAG,GAG |
K101E |
XP_016873152.1 |
XM_017017664.1 |
434 |
Missense Mutation |
AAG,GAG |
K137E |
XP_016873153.1 |
XM_017017665.1 |
434 |
Missense Mutation |
AAG,GAG |
K137E |
XP_016873154.1 |
XM_017017666.1 |
434 |
Missense Mutation |
AAG,GAG |
K87E |
XP_016873155.1 |
XM_017017667.1 |
434 |
Missense Mutation |
AAG,GAG |
K101E |
XP_016873156.1 |
XM_017017668.1 |
434 |
Missense Mutation |
AAG,GAG |
K87E |
XP_016873157.1 |
- Gene
- TALDO1
- Gene Name
- transaldolase 1
There are no transcripts associated with this gene.
View Full Product Details