Product Details

SNP ID
rs150250361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5199802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTGTTGGGTCTTGACACTGTAG[A/G]TGATGGGATTCATTAAAGGTGGGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51V1 PubMed Links

Gene Details

Gene
OR51V1
Gene Name
olfactory receptor family 51 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004760.2 899 Missense Mutation ACC,ATC T300I NP_001004760.2

View Full Product Details