Product Details

SNP ID
rs150295666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5884368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAATACCAGGACTGGACACTTTA[A/C]ATATCTGGATTTCTTTCCCATTCTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR52E4 PubMed Links

Gene Details

Gene
OR52E4
Gene Name
olfactory receptor family 52 subfamily E member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005165.1 76 Missense Mutation AAT,CAT N26H NP_001005165.1

View Full Product Details