Product Details

SNP ID
rs150503343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74268169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCACTTATCTCCCACCAGGACAG[A/G]ACAGCCAGCATGAAGTGTGTCACTG
Phenotype
MIM: 608987
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
P4HA3 PubMed Links

Gene Details

Gene
P4HA3
Gene Name
prolyl 4-hydroxylase subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288748.1 1605 Silent Mutation GTC,GTT V511V NP_001275677.1
NM_182904.4 1605 Missense Mutation CCT,TCT P514S NP_878907.1

View Full Product Details