Product Details
- SNP ID
-
rs150960595
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:110435891 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCGTGATGGTGAAACATTAACAAC[C/T]AAAGGAAAAGTTGGTGATTCTCTGC
- Phenotype
-
MIM: 103260
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FDX1
PubMed Links
Gene Details
- Gene
- FDX1
- Gene Name
- ferredoxin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004109.4 |
427 |
Silent Mutation |
ACC,ACT |
T81T |
NP_004100.1 |
View Full Product Details