Product Details

SNP ID
rs151023406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118349182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGGGGATAGAGAGGTGATGTCTC[C/T]ATTGTCAACCAAATCAGTGACCTGA
Phenotype
MIM: 186790 MIM: 186740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD3D PubMed Links

Gene Details

Gene
CD3D
Gene Name
CD3d molecule
There are no transcripts associated with this gene.

Gene
CD3G
Gene Name
CD3g molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000073.2 Intron NP_000064.1
XM_005271724.3 Intron XP_005271781.1
XM_006718941.2 Intron XP_006719004.1

View Full Product Details