Product Details

SNP ID
rs151191536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5200118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCGGATGTCTGAACAGGCTAAGC[A/G]GAGAAGATCCTGGTGCAGGCAGAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51V1 PubMed Links

Gene Details

Gene
OR51V1
Gene Name
olfactory receptor family 51 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004760.2 583 Missense Mutation CGC,TGC R195C NP_001004760.2

View Full Product Details