Product Details

SNP ID
rs138102893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55130190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTGACTGAGGGATTTCTCCTGG[C/T]GGCCATGGCTTATGACCGCTTTATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR9K2 PubMed Links

Gene Details

Gene
OR9K2
Gene Name
olfactory receptor family 9 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005243.1 422 Missense Mutation GCG,GTG A141V NP_001005243.1

View Full Product Details