Product Details
- SNP ID
-
rs138269355
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:123606119 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCGCTTTTGTCATCCCCATCCTGG[A/G]AATTCTTCTGAGAAGAGAAGAGAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DDX55
PubMed Links
Gene Details
- Gene
- DDX55
- Gene Name
- DEAD-box helicase 55
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020936.2 |
219 |
Missense Mutation |
GAA,GGA |
E69G |
NP_065987.1 |
XM_011538608.2 |
219 |
Missense Mutation |
GAA,GGA |
E69G |
XP_011536910.1 |
XM_017019710.1 |
219 |
Missense Mutation |
GAA,GGA |
E69G |
XP_016875199.1 |
XM_017019711.1 |
219 |
Missense Mutation |
GAA,GGA |
E31G |
XP_016875200.1 |
XM_017019712.1 |
219 |
Missense Mutation |
GAA,GGA |
E26G |
XP_016875201.1 |
XM_017019713.1 |
219 |
Missense Mutation |
GAA,GGA |
E26G |
XP_016875202.1 |
XM_017019714.1 |
219 |
Missense Mutation |
GAA,GGA |
E26G |
XP_016875203.1 |
XM_017019715.1 |
219 |
Missense Mutation |
GAA,GGA |
E69G |
XP_016875204.1 |
XM_017019716.1 |
219 |
Missense Mutation |
GAA,GGA |
E69G |
XP_016875205.1 |
XM_017019717.1 |
219 |
UTR 5 |
|
|
XP_016875206.1 |
XM_017019718.1 |
219 |
UTR 5 |
|
|
XP_016875207.1 |
XM_017019719.1 |
219 |
UTR 5 |
|
|
XP_016875208.1 |
XM_017019720.1 |
219 |
UTR 5 |
|
|
XP_016875209.1 |
XM_017019721.1 |
219 |
Missense Mutation |
GAA,GGA |
E69G |
XP_016875210.1 |
- Gene
- TMED2
- Gene Name
- transmembrane p24 trafficking protein 2
There are no transcripts associated with this gene.
View Full Product Details