Product Details

SNP ID
rs138477792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54608990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTAACATACAAGTAGGACTGAG[A/G]TCCCGTATCAGAATCCAAATCTATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLYCAM1 PubMed Links

Gene Details

Gene
GLYCAM1
Gene Name
glycosylation dependent cell adhesion molecule 1 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details