Product Details

SNP ID
rs138478563
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:101479614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTACCTGGCTTTAATCAACCAT[C/T]GTCCTCATGTCAAAGGAAATTCCAG
Phenotype
MIM: 612568
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SPIC PubMed Links

Gene Details

Gene
SPIC
Gene Name
Spi-C transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005268646.3 Intron XP_005268703.1
XM_006719239.3 Intron XP_006719302.1

View Full Product Details