Product Details

SNP ID
rs138497401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48693154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCGAGGATTCAGATAGTCACTA[C/T]AAGGACGAACAAATTCAAATGCAGT
Phenotype
MIM: 143055
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNT1 PubMed Links

Gene Details

Gene
CCNT1
Gene Name
cyclin T1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001240.3 2219 Missense Mutation TAT,TGT Y687C NP_001231.2
NM_001277842.1 2219 UTR 3 NP_001264771.1
XM_017020197.1 2219 Missense Mutation TAT,TGT Y607C XP_016875686.1

View Full Product Details