Product Details

SNP ID
rs138524511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:42459866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTTCTTCTTGGATTTTGTTGT[C/T]CTCTGACCAAACTGAGGGGTGGGAA
Phenotype
MIM: 608500
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRICKLE1 PubMed Links

Gene Details

Gene
PRICKLE1
Gene Name
prickle planar cell polarity protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144881.1 2544 Silent Mutation AGA,AGG R813R NP_001138353.1
NM_001144882.1 2544 Silent Mutation AGA,AGG R813R NP_001138354.1
NM_001144883.1 2544 Silent Mutation AGA,AGG R813R NP_001138355.1
NM_153026.2 2544 Silent Mutation AGA,AGG R813R NP_694571.2
XM_011537946.1 2544 Silent Mutation AGA,AGG R813R XP_011536248.1
XM_011537947.2 2544 Silent Mutation AGA,AGG R813R XP_011536249.1
XM_017018838.1 2544 Silent Mutation AGA,AGG R813R XP_016874327.1
XM_017018839.1 2544 Silent Mutation AGA,AGG R813R XP_016874328.1
XM_017018840.1 2544 Silent Mutation AGA,AGG R813R XP_016874329.1

View Full Product Details