Product Details

SNP ID
rs138760241
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:82354522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCTTAGCAGCACGTTTGGCTTG[A/T]ATCTGTTCATATTCTTCTTGTGCTT
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
CCDC59 PubMed Links

Gene Details

Gene
CCDC59
Gene Name
coiled-coil domain containing 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014167.4 581 Silent Mutation ATA,ATT I179I NP_054886.2
Gene
METTL25
Gene Name
methyltransferase like 25
There are no transcripts associated with this gene.

View Full Product Details