Product Details
- SNP ID
-
rs139024217
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:123584673 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGAACTGCTGGTGCTTCTGGCC[A/G]CTCTCCTGGCCACGGTCTCGGGCTA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101927415
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11551723] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101927415
- Gene Name
- uncharacterized LOC101927415
There are no transcripts associated with this gene.
- Gene
- TMED2
- Gene Name
- transmembrane p24 trafficking protein 2
View Full Product Details