Product Details

SNP ID
rs139105553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55130170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCTCTTTGCCCTCCTCATTGT[A/G]ACTGAGGGATTTCTCCTGGCGGCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR9K2 PubMed Links

Gene Details

Gene
OR9K2
Gene Name
olfactory receptor family 9 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005243.1 402 Silent Mutation GTA,GTG V134V NP_001005243.1

View Full Product Details