Product Details

SNP ID
rs139443241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53308073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCAGGGAAGGAGCTCAAACACA[C/G]GGCTGTTTCGAGTGCGAAAAAGGAG
Phenotype
MIM: 605378 MIM: 611366 MIM: 604899
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AAAS PubMed Links

Gene Details

Gene
AAAS
Gene Name
aladin WD repeat nucleoporin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173466.1 1405 Missense Mutation CCT,CGT P404R NP_001166937.1
NM_015665.5 1405 Missense Mutation CCT,CGT P437R NP_056480.1
XM_011538778.1 1405 Missense Mutation CCT,CGT P437R XP_011537080.1
XM_011538780.1 1405 Missense Mutation CCT,CGT P404R XP_011537082.1
Gene
C12orf10
Gene Name
chromosome 12 open reading frame 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021640.3 1405 Intron NP_067653.3
Gene
PFDN5
Gene Name
prefoldin subunit 5
There are no transcripts associated with this gene.

View Full Product Details