Product Details

SNP ID
rs139500653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48525701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCTGTCTGCCGACCCCAACATC[C/T]GGGCTCTGCTCTTTGTGCTGTTCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR8S1 PubMed Links

Gene Details

Gene
OR8S1
Gene Name
olfactory receptor family 8 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005203.2 70 Missense Mutation CGG,TGG R24W NP_001005203.2

View Full Product Details