Product Details
- SNP ID
-
rs139525088
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:19129829 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCGGATCTGAACCTGGAGTGGAT[C/T]TCCCTGCCCCGGTCCTGGACTTACG
- Phenotype
-
MIM: 607770
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLEKHA5
PubMed Links
Gene Details
- Gene
- PLEKHA5
- Gene Name
- pleckstrin homology domain containing A5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001143821.2 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
NP_001137293.2 |
NM_001190860.2 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
NP_001177789.2 |
NM_001256470.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
NP_001243399.1 |
NM_001256787.1 |
98 |
Intron |
|
|
NP_001243716.1 |
NM_019012.5 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
NP_061885.2 |
XM_005253400.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_005253457.1 |
XM_006719091.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_006719154.1 |
XM_006719093.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_006719156.1 |
XM_006719096.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_006719159.1 |
XM_011520714.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_011519016.1 |
XM_011520715.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_011519017.1 |
XM_011520716.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_011519018.1 |
XM_011520717.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_011519019.1 |
XM_017019495.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_016874984.1 |
XM_017019496.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_016874985.1 |
XM_017019497.1 |
98 |
Intron |
|
|
XP_016874986.1 |
XM_017019498.1 |
98 |
Intron |
|
|
XP_016874987.1 |
XM_017019499.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_016874988.1 |
XM_017019500.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_016874989.1 |
XM_017019501.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_016874990.1 |
XM_017019502.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_016874991.1 |
XM_017019503.1 |
98 |
Silent Mutation |
ATC,ATT |
I10I |
XP_016874992.1 |
View Full Product Details