Product Details

SNP ID
rs139716647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108623095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGGAGGAAGCTGTGCAGGGTGA[A/G]GTCATCCCCCTCACGGTCCTCCCTG
Phenotype
MIM: 600738
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SELPLG PubMed Links

Gene Details

Gene
SELPLG
Gene Name
selectin P ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206609.1 1438 Missense Mutation CTC,TTC L421F NP_001193538.1
NM_003006.4 1438 Missense Mutation CTC,TTC L405F NP_002997.2

View Full Product Details