Product Details

SNP ID
rs139903785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10702119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAACTGCTGGGGCAGGTCGTGG[A/G]CGAGGAGGTCCCCTGCTGTAGGGAA
Phenotype
MIM: 603437
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
YBX3 PubMed Links

Gene Details

Gene
YBX3
Gene Name
Y-box binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145426.1 Intron NP_001138898.1
NM_003651.4 Intron NP_003642.3
XM_011520871.2 Intron XP_011519173.1
XM_017020122.1 Intron XP_016875611.1
XM_017020123.1 Intron XP_016875612.1

View Full Product Details