Product Details
- SNP ID
-
rs139961528
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:6541629 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCCGGTTCATGTCGTTCTTGGCC[A/G]TGGCCAACTCCAGCTGTGGTGGGGC
- Phenotype
-
MIM: 138400
MIM: 610495
MIM: 615638
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAPDH
PubMed Links
Gene Details
- Gene
- GAPDH
- Gene Name
- glyceraldehyde-3-phosphate dehydrogenase
There are no transcripts associated with this gene.
- Gene
- IFFO1
- Gene Name
- intermediate filament family orphan 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001039670.2 |
1806 |
Missense Mutation |
ACG,ATG |
T490M |
NP_001034759.1 |
NM_001193457.1 |
1806 |
Missense Mutation |
ACG,ATG |
T498M |
NP_001180386.1 |
NM_080730.4 |
1806 |
Missense Mutation |
ACG,ATG |
T489M |
NP_542768.2 |
XM_006718975.3 |
1806 |
Missense Mutation |
ACG,ATG |
T489M |
XP_006719038.1 |
XM_006718976.3 |
1806 |
Missense Mutation |
ACG,ATG |
T487M |
XP_006719039.1 |
XM_006718977.3 |
1806 |
Missense Mutation |
ACG,ATG |
T486M |
XP_006719040.1 |
XM_006718981.3 |
1806 |
Missense Mutation |
ACG,ATG |
T182M |
XP_006719044.1 |
XM_011520947.2 |
1806 |
Missense Mutation |
ACG,ATG |
T578M |
XP_011519249.1 |
XM_011520948.2 |
1806 |
Missense Mutation |
ACG,ATG |
T577M |
XP_011519250.1 |
XM_011520949.2 |
1806 |
Missense Mutation |
ACG,ATG |
T575M |
XP_011519251.1 |
XM_011520952.2 |
1806 |
Missense Mutation |
ACG,ATG |
T270M |
XP_011519254.1 |
XM_017019169.1 |
1806 |
Intron |
|
|
XP_016874658.1 |
XM_017019170.1 |
1806 |
Missense Mutation |
ACG,ATG |
T515M |
XP_016874659.1 |
XM_017019171.1 |
1806 |
Missense Mutation |
ACG,ATG |
T488M |
XP_016874660.1 |
XM_017019172.1 |
1806 |
Missense Mutation |
ACG,ATG |
T271M |
XP_016874661.1 |
XM_017019173.1 |
1806 |
Missense Mutation |
ACG,ATG |
T268M |
XP_016874662.1 |
- Gene
- NCAPD2
- Gene Name
- non-SMC condensin I complex subunit D2
There are no transcripts associated with this gene.
View Full Product Details