Product Details

SNP ID
rs140213362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:14978038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGTGACAACACTACTCTGCCTG[C/T]TCCAGCTTCAAACCAGGGTCCTACC
Phenotype
MIM: 601190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE6H PubMed Links

Gene Details

Gene
PDE6H
Gene Name
phosphodiesterase 6H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006205.2 375 Missense Mutation GCT,GTT A9V NP_006196.1
XM_017019431.1 375 Missense Mutation GCT,GTT A9V XP_016874920.1

View Full Product Details