Product Details

SNP ID
rs140344263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:22046499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCCCCTGAAGAACATTAAGCAC[C/T]TGGCGGGGGTCCCGCTCATTGGCTG
Phenotype
MIM: 603316
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CMAS PubMed Links

Gene Details

Gene
CMAS
Gene Name
cytidine monophosphate N-acetylneuraminic acid synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018686.5 326 Silent Mutation CTG,TTG L66L NP_061156.1

View Full Product Details