Product Details

SNP ID
rs140778366
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55819431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGCTACTGACACACTGGGAGCA[A/G]ATGGACTATGGGCTCCAGTTTACCT
Phenotype
MIM: 606092 MIM: 610074 MIM: 610049
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DNAJC14 PubMed Links
Additional Information
For this assay, SNP(s) [rs56108400] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DNAJC14
Gene Name
DnaJ heat shock protein family (Hsp40) member C14
There are no transcripts associated with this gene.

Gene
ORMDL2
Gene Name
ORMDL sphingolipid biosynthesis regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014182.4 356 Silent Mutation CAA,CAG Q88Q NP_054901.1
Gene
SARNP
Gene Name
SAP domain containing ribonucleoprotein
There are no transcripts associated with this gene.

View Full Product Details